missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers
POP-7™ Polymer is a separation matrix for the 3730 and 3730xl DNA Analyzers. POP-7™ Polymer is customized for DNA sequencing and fragment analysis.
£1326.00 - £11330.00
Specifications
Accuracy | 98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary) |
---|---|
For Use With (Application) | Standard Sequencing, Fragment Analysis (Denaturing) |
Product Line | POP-7™ |
Product Type | Polymer |
Test Time | 1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.) |
Includes: Each bottle contains enough polymer for 960 samples
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15829816
|
Applied Biosystems™
4363929 |
1 x 28 mL |
£1682.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15781816
|
Applied Biosystems™
4335615 |
5 x 28 mL |
£3820.00
Pack of 5 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15839816
|
Applied Biosystems™
4363935 |
10 x 28 mL |
£5020.00
Each |
In Stock
Log in to see stock. |
|||||
15869776
|
Applied Biosystems™
4335611 |
30 x 28 mL |
£11330.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16375302
|
Applied Biosystems™
4363929 |
1 x 28 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919182
|
Applied Biosystems™
4335615 |
5 x 28 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- A separation matrix for the 3730 and 3730xl DNA Analyzers
- Customized for DNA sequencing and fragment analysis.
- Pre-formulated to save time and reproducibility by ensuring consistent formulation
- Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
- Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
- Sufficient for 8,000 to 12,000 samples
Use With
- 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
- 3730 Running Buffer, 10X
- BigDye™ Terminator v1.1 Cycle Sequencing Kit
- BigDye™ Terminator v3.1 Cycle Sequencing Kit
- Hi-Di™ Formamide
Specifications
98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary) | |
POP-7™ | |
1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.) |
Standard Sequencing, Fragment Analysis (Denaturing) | |
Polymer |
For Research Use Only. Not for use in diagnostic procedures.