missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | POP7 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863904
|
Proteintech
14964-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853904
|
Proteintech
14964-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP complex, which cleaves pre-rRNA sequences.Specifications
POP7 | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75817 | |
10248 | |
POP7 Fusion Protein Ag6827 | |
Primary | |
-20°C | |
POP7 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
0610037N12Rik, hPOP7, POP7, RNaseP protein p20, RPP2, RPP20 | |
POP7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |