Learn More
Haldenwanger™ Porcelain Buchner Funnel
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
Brand:  Haldenwanger™ 127C/1
 View more versions of this product
Product Code. 10343221
  • £174.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
  • Vertical sides
  • Available in a variety of sizes to fit many filtration needs
  • Perforated fixed filter plate
Specifications
Specifications
75 mL
59mm
Buchner Funnel
Porcelain
1Pack