missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Casseroles
£15.54 - £64.05
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10751562
|
Haldenwanger™
17/2 |
500 mL |
£63.30
Each |
In Stock
Log in to see stock. |
|||||
10220961
|
Haldenwanger™
17/0 |
220 mL |
£15.54
Each |
In Stock
Log in to see stock. |
|||||
10701392
|
Haldenwanger™
17/000 |
140 mL | N/A | ||||||
Specifications
Porcelain | |
Round | |
Casserole |