Learn More
Haldenwanger™ Porcelain Pestles
Use these porcelain pestles for a wide range of applications. Haldenwanger™ Porcelain Pestles are available glazed or unglazed with head sizes from 24 to 85mm in diameter. Stringent inspections, starting with raw materials and continuing down to finished products, guarantee constant quality.
Brand:  Haldenwanger™ 56/00
 View more versions of this product
Product Code. 10405011
  • £25.70 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Grinding surfaces are unglazed
  • Excellent chemical resistance
  • Laboratory porcelain complies with DIN EN 60672-3,Group C100, Type C110
Specifications
Specifications
Unglazed
Pestle
115mm
24mm
Porcelain