Learn More
R&D Systems™ Porcine C-Reactive Protein/CRP DuoSet ELISA
Porcine C-Reactive Protein/CRP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY2648
This item is not returnable.
View return policy
Description
The Porcine C-Reactive Protein/CRP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies porcine C-Reactive Protein/CRP.Specifications
Biotin | |
78.10 to 5000 pg/mL | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |