missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Porcine IL-10 DuoSet ELISA
Porcine IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY693B
Additional Details : Weight : 0.09700kg
Description
The Porcine IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies porcine IL-10.Specifications
Biotin | |
ELISA | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
23.40 to 1500 pg/mL | |
397106 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |