missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Porcine Serum
GREENER_CHOICE

Product Code. 11500646
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500646

Brand: Gibco™ 26250084

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Standard (sterile-filtered)

  • Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • Sera Category: Specialty
  • Origin: New Zealand
  • Age: 1 year or less

Order Info

Shipping condition: Frozen (Dry Ice)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Swine/Porcine, Porcine
Age 1 year or less
Country of Origin New Zealand
Form Liquid
Product Type Porcine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Product Line Gibco
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.