Learn More
Thermo Scientific™ POROS™ GoPure™ Benzyl Ultra Pre-packed Column, 0.8 x 10 cm, 5 mL
POROS GoPure 0.8 cm pre-packed columns are intended for the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) and hydrophobic interaction chromatography (HIC) resins or for small-scale purification.
Brand: Thermo Scientific™ A36655
Additional Details : CAS Number : 64-17-5 Weight : 0.10000kg
Description
POROS GoPure 0.8 cm pre-packed columns are intended for the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) and hydrophobic interaction chromatography (HIC) resins or for small-scale purification. These pre-packed columns are ready to connect and have the following features:
- Dimensions of 0.8 x 10 cm, with a nominal bed volume of 5 mL, providing a uniform screening platform
- Direct connection to standard chromatography systems
- Convenient—ready-to-run columns simplify and speed POROS resin evaluations
- The 10-cm bed height and 5-mL bed volume allow initial process development at a bench scale
- Packed with POROS high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS Benzyl Ultra HIC resin (Cat. No. A32564) in this GoPure column has an aromatic hydrophobic benzyl ligand, well suited for hydrophobic interaction chromatography (HIC) for aggregate removal from monoclonal antibody monomer in flow-through mode at low salt concentrations.
Other POROS hydrophobic interaction and ion exchange resins are also available in this column format. All POROS High Performance Chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
Specifications
5 mL | |
Benzyl Ultra Resin | |
Room Temperature | |
GoPure™, POROS™ |
0.8 cm | |
1 Column | |
Chromatography Column | |
10 cm |