Learn More
Thermo Scientific™ POROS™ GoPure™ Ethyl Pre-packed Column, 0.5 x 5 cm, 1 mL
POROS GoPure 0.5 cm pre-packed columns are intended for the screening of POROS ion exchange and hydrophobic interaction chromatography (HIC) resins or for small-scale purification.
Brand: Thermo Scientific™ A34983
79.33 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : CAS Number : 64-17-5 Weight : 0.25000kg
Description
POROS GoPure 0.5 cm pre-packed columns are intended for the screening of POROS ion exchange and hydrophobic interaction chromatography (HIC) resins or for small-scale purification. These pre-packed columns are ready to connect and have the following features:
• Dimensions of 0.5 x 5 cm, with a nominal bed volume of 1 mL, providing a uniform screening platform
• Direct connection to standard chromatography systems
• Packed with POROS high-performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS Ethyl HIC Resin (Cat. No. A32552) in this GoPure column has a novel hydrophobic ethyl ligand, making it suited primarily for bind/elute purifications.
- Convenient—ready-to-run columns simplify and speed POROS resin evaluations
- Conserves sample—the 1 mL nominal bed volume minimizes sample requirement for screening multiple resins
- Simplified scalability—the consistency and range of the POROS GoPure family of pre-packed columns simplifies process scale-up and process modeling
POROS GoPure 0.5 cm pre-packed columns have been developed to meet the needs of the process development scientist requiring a convenient and uniform POROS resin-screening platform. The 1 mL bed volume reduces sample requirements, and the recommended >3 min residence time (0.3 mL/min) on this 5-cm-length column equates to 400 cm/hr on a 20-cm bed height. Detailed specifications may be found in the POROS GoPure 0.5 cm Operating Instructions below.
Other POROS hydrophobic interaction and ion exchange resins are also available in this column format. All POROS High Performance Chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
For scaleup, POROS GoPure pre-packed columns are available in a range of diameters and in multiple bed heights that equate to a column volume range of 5 mL to 22 mL.
Specifications
0.33 mL/min. | |
Room Temperature | |
Novel Ethyl | |
1 mL | |
POROS™ |
Store at room temperature. | |
Pre-packed Column | |
0.5 cm | |
1 Column | |
5 cm |