Learn More
Thermo Scientific™ POROS™ GoPure™ Ethyl Pre-packed Column, 0.8 x 10 cm, 5 mL
POROS GoPure 0.8 cm pre-packed columns are intended for the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) and hydrophobic interaction chromatography (HIC) resins or for small-scale purification.
Brand: Thermo Scientific™ A36653
Additional Details : CAS Number : 64-17-5 Weight : 0.10000kg
Description
POROS GoPure 0.8 cm pre-packed columns are intended for the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) and hydrophobic interaction chromatography (HIC) resins or for small-scale purification. These pre-packed columns are ready to connect and have the following features:
- Dimensions of 0.8 x 10 cm, with a nominal bed volume of 5 mL, providing a uniform screening platform
- Direct connection to standard chromatography systems
- Convenient—ready-to-run columns simplify and speed POROS resin evaluations
- The 10-cm bed height and 5-mL bed volume allow initial process development at a bench scale
- Packed with POROS high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS Ethyl HIC resin (Cat. No. A32552) in this GoPure column has a novel hydrophobic ethyl ligand, making it suited primarily for bind/elute purifications.
Other POROS hydrophobic interaction and ion exchange resins are also available in this column format. All POROS High Performance Chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
Specifications
5 mL | |
Ethyl Resin | |
Room Temperature | |
GoPure™, POROS™ |
0.8 cm | |
1 Column | |
Chromatography Column | |
10 cm |