Learn More
Thermo Scientific™ POROS™ GoPure™ XQ Pre-packed Column, 0.5 x 5 cm, 1 mL
POROS™ GoPure™ pre-packed columns are intended for screening of POROS™ ion exchange resins or for small scale purification.
Brand: Thermo Scientific™ A25812
79.33 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : CAS Number : 64-17-5 Weight : 0.10000kg
Description
POROS™ GoPure™ pre-packed columns are intended for screening of POROS™ ion exchange resins or for small scale purification. These prepacked columns are ready to connect and have the following features:
- Dimensions of 0.5 x 5 with a nominal 1 mL bed volume that provides a uniform screening platform
- Direct connections to standard chromatography systems
- Packed with POROS™ high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS™ XQ resin in this GoPure™ column features a strong anion exchange functional group and is a high capacity and high resolution resin with excellent salt tolerance.
- Convenient—ready-to-run columns simplify and speed POROS™ resin evaluations
- Conserves sample—GoPure™ 0.5 x 5 cm columns, with their 1 mL nominal bed volume, minimize the sample needed for screening multiple resins
- Simplified scalability—reproducible GoPure™ pre-packed columns simplify process scale up or process modeling
GoPure™ 0.5 x 5 cm pre-packed columns have been developed to meet the needs of the process development scientist requiring a convenient and uniform POROS™ resin screening platform. The 1 mL bed volume reduces sample requirements, and a recommended ≥3 minute residence time (0.3 mL/min.) on this 5 cm length column equates to 400 cm/hr on a 20 cm bed height.
Other POROS™ anion exchange resins, as well cation exchange resins, are also available in this column format. All POROS™ high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
For scale-up, POROS™ GoPure™ pre-packed columns are available in a range of diameters and in multiple bed heights that equate to a column volume range of 5 mL to 6.3 L.
Specifications
0.33 mL/min. | |
Room Temperature | |
Proprietary Strong Anion Exchange | |
1 mL | |
GoPure™ |
>140 mg/mL (In the presence of up to 6.6 mS/cm NaCl) | |
Anion Exchange | |
0.5 cm | |
1 Column | |
5 cm |