Learn More
Thermo Scientific™ POROS™ GoPure™ XQ Pre-packed Column, 0.8 x 10 cm, 5 mL
POROS GoPure 0.5 x 5 cm prepacked columns are intended for screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) resins or for small scale purification.
Brand: Thermo Scientific™ A36640
441.35 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : CAS Number : 64-17-5 Weight : 0.10000kg
Description
POROS GoPure 0.5 x 5 cm prepacked columns are intended for screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) resins or for small scale purification. These prepacked columns are ready to connect and have the following features:
- Dimensions of 0.8 cm x 10 cm, with a nominal bed volume of 5 mL, providing a uniform screening platform
- Direct connection to standard chromatography systems
- Convenient—ready-to-run columns simplify and speed POROS resin evaluations
- The 10-cm bed height and 5-mL bed volume allow initial process development at a bench scale
- Packed with POROS high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS XQ resin (Cat. No. 82073) in this GoPure column features a strong anion exchange functional group and is a high capacity and high resolution resin with excellent salt tolerance.
Other POROS anion exchange resins, as well cation exchange resins, are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
Specifications
5 mL | |
Small Scale Purification, Strong Anion Exchange | |
POROS GoPure | |
POROS XQ Resin | |
100 mm |
Proprietary Quaternary Amine | |
435 psi (30 bar) | |
Room Temperature | |
Pre-packed Column | |
0.8 mm |