Learn More
Fisherbrand™ Portable Balance
Portable Balance with a slim, stackable design provides basic weighing functionality and competitive performance.
Brand:  Fisherbrand™ 30467786
29.87 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 15949345
  • £59.05 / Each
Explore more special offers
Description
Description
  • Intuitive two-button design makes for easy operation
  • Slim, space-saving design allows for stackable storage
  • Enlarged weighing platform can adequately handle large samples and a variety of objects
TRUSTED_SUSTAINABILITY
Specifications
Specifications
620 g
0.100000 g
3 AA (LR6) Batteries
2.000 sec.
18°C to 25°C
Gram, Pound, Ounce, Ounce Troy, Pennyweight
5 in.
5.2 in.
0.9 lb.
LCD
Weighing
FCS621
64°F to 77°F
2 Years
205 x 140 x 41 mm (8.1 x 5.5 x 1.6 in.)
125 mm
132 mm
0.4 kg