Learn More
KNF™ Portable Diaphragm Vacuum Pump
Use this pump for sampling, evacuating and compressing applications. KNF™ Portable Diaphragm Vacuum Pumps permit gentle distillation and high rates of recovery even with low-boiling solvents.
Brand: KNF™ 046209/049540
458.83 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Small portable diaphragm pumps, ideal for general use in the laboratory and educational establishments
- Compact and portable
- Chemically resistant
- Integrated retractable handle
Specifications
160 mbar | |
Ryton head, PTFE coated diaphragm and Kalrez valves. | |
5.5 L/min. | |
141 mm | |
Vacuum Pump | |
230 V |
90 mm | |
1.9 kg | |
50 Hz | |
164 mm | |
160 mbar |