Learn More
Esculape Laboratoire™ Portable Diphoterine Shower
Rechargeable and ready to use portable autonomous Diphoterine shower to neutralize chemical burns on the hands, face or body.
Brand:  Esculape Laboratoire™ 8205263
Product Code. 15718079
  • £1360.00 / Each
Explore more special offers
Description
Description
  • Easily identifiable with its green color - differentiates from a fire extinguisher
  • No specific training required
  • Ready to use, allows rapid action
  • Soft, micronized and very wetting jet allowing washing without trauma
  • Long hose allows total and complete washing of the body
  • Sterile solution
  • No special maintenance and no special installation
  • Rechargeable
  • Complies with EN 15154-3 - CE approved
Specifications
Specifications
Green
EN 15154-3, CE approved
640 x 220 x 200 mm
5 L Sterile Solution to Neutralize and Decontaminate Chemical Burns and Splashes. 7 Bar Pressure with CO2 Propellant Gas