missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters
Designed to measure pH, conductivity (EC), total dissolved solids (TDS), and temperature with a single probe. Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters are available for high range and low range measurement.
£1135.00
Specifications
Accuracy (pH) | ±0.01 |
---|---|
Accuracy (Temperature) | ±0.5°C/±1°F |
Includes | HI 1288 probe, batteries, 1m cable, rugged carrying case and instructions |
Calibration | Automatic |
Range (pH) | 0 to 14 |
Product Code | Brand | For Use With (Application) | Range (Conductivity) | Range (TDS) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Range (Conductivity) | Range (TDS) | Price | Quantity & Availability | |||
10142321
|
Hanna Instruments™
HI991301 |
High-Range Detection | 0.00 to 20.00mS/cm | 0.00 to 10.00ppt (g/L) |
£1135.00
Each |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
This multi-parameter meter takes readings simply and quickly, and it fits in the palm of your hand. From setup to calibration all operations are performed with just two buttons. It features a clear, extra-large LCD display and easy-access rotary knobs to effect calibration changes. A single pre-amplified probe is used for all parameters.
- Waterproof and rated for IP 67 conditions
- Easy to clean and maintain
- Multi-level LCD display
- On-screen tutorial messages for calibration and setup
- Automatic Temperature Compensation
- Automatic one or two-point pH calibration
- BEPS: Alerts the user in the event that low battery power could adversely affect the readings
- Battery % displayed on startup
- Compact, heavy duty, and waterproof
- Preprogrammed buffer recognition values, TDS factors and temperature coefficients
Specifications
±0.01 | |
HI 1288 probe, batteries, 1m cable, rugged carrying case and instructions | |
0 to 14 | |
0°C to 60.0°C | |
0.1°C/F | |
IP 67 | |
205 g | |
2 Years (6 Months Probe) | |
±2% F.S. | |
HI1288 PVC body, pre-amplified multiparameter probe with internal temperature sensor, DIN connector and 1m cable | |
0 to 14 pH | |
1.5V (3) AAA, Auto-Off after 8 min. non-use | |
500 hr. |
±0.5°C/±1°F | |
Automatic | |
0.01 | |
32.0°F to 140.0°F | |
LCD | |
150 x 80 x 36 mm (5.9 x 3.1 x 1.4 in.) | |
7.2 oz. | |
Two-button | |
PVC | |
TDS Conversion Factor: 0.45 to 1.00 | |
pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4%/°C | |
Portable pH/EC/TDS/Temperature Meter | |
±2% full scale |