missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Portable Photometer
The portable pHotoFlex™ STD with energy-efficient LED optics is equipped with 6 wavelengths and a foldable adapter for various applications
Brand: WTW™ 251105
1619.28 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The portable pHotoFlex™ STD with energy-efficient LED optics is equipped with 6 wavelengths and a foldable adapter for various applications with 16 mm and 28 mm cuvettes, for even the smallest concentrations. With more than 180 programs for standard parameters and coloration as well as a multitude of test kits - from lot-certified cuvettes to economic powder reagents, it is suitable for monitoring by health offices via environmental monitoring all the way to service labs. A list of favorites for the most commonly used tests, sample ID numbers and more than 3,000 measurements per battery set facilitate field measurements. An elegant folding mechanism allows the easy switch between the two cuvette sizes without the need to exchange the cuvette holder. The optional PC software LSdata offers GLP-compliant data evaluation with data filters and the input of up to 10 user-defined programs, calculate them as a method automatically and save them to pHotoFlex™. For convenient work in the field, a case set is a available as a mobile lab with a work surface and storage compartment as well as a lot of accessories, such as pipettes and cuvettes and spare batteries. For lab use, the photometer can be used in conjunction with a LabStation (optional) with site-supplied power and fitted with an external barcode reader.
Specifications
Portable Photometer | |
LED | |
1.5 V | |
LED |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Portable Photometer > Product Type: Portable Photometer
Spot an opportunity for improvement?Share a Content Correction