missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Portable Precision Balance - NVT Series

Multi-Purpose Portable Balances Suitable for Everyday Weighing

Brand:  OHAUS™ 30467836

259.35 GBP valid until 2024-12-31
Use promo code "21987" to get your promotional price.



Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 16660314

  • £273.00 / Each
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
Specifications

Specifications

RS232, USB, or Ethernet (available as accessories)
3200.00 g
1.0000 g
1.0000 g
1.0000 g
Gram, Kilogram, & Carat
9.80 in.
9.50 in.
240.00 mm
2.90 in.
9.80 in.
250.00 mm
3.300 lb.
AC adapter (included) or 4 C batteries (not included)
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Backlit LCD
Weighing, Parts Counting, Percent Weighing, Checkweighing
NVT3200M
1.000 s
39.000 x 32.500 x 17.000 cm
250.00 mm
9.100 in.
230.000 mm
74.000 mm
6.900 in.
174.000 mm
1.500 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers