missing translation for 'onlineSavingsMsg'
Learn More
Posaconazol, Honeywell Fluka™ Product Code.: 15941151

Posaconazol, Honeywell Fluka™

Product Code. 15941151 Shop All Honeywell Fluka Products
25 mg
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy

Product Code. 15941151

Brand: Honeywell Fluka R131625MG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 171228-49-2
Packaging Glass Bottle
Quantity 25 mg
For Use With (Application) Antibacterial Agents
Physical Form Powder
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Posaconazol, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.