Learn More
Gilson™ Position Rack Flask Vessel Type
The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
Brand:  Gilson™ 152424
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16636125
  • £1102.40 / Pack of 5
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
  • These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
  • Flask Vessel Type
Specifications
Specifications
Code 22
25 mL Vial (18 x 150 mm)
5 Pk.
Flask
44