missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Position Rack Flask Vessel Type

The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates

Brand:  Gilson™ 150498

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 16606125

  • £407.04 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
  • Flask Vessel Type
Specifications

Specifications

Code 22 U
Flasks
44
10 to 18 mm x 100 to 175 mm Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers