Learn More
Gilson™ Position Rack Flask Vessel Type
The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
Brand:  Gilson™ 2704341
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16616155
  • £486.54 / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
  • These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
  • Flask Vessel Type
Specifications
Specifications
Code 29 SE
Flasks
60
60 x 0.5 mL Microcentrifuge Tubes