Learn More
Gilson™ Position Rack Flask Vessel Type
The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
Brand: Gilson™ 150429
Additional Details : Weight : 0.45000kg
Description
- These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
- Flask Vessel Type
Specifications
Code 29 | |
Flasks | |
60 | |
60 Tubes (12/13 x 75 mm) |