Learn More
Gilson™ Position Rack Tube Vessel Type, Fraction Collector
These racks are for use with the FC203B and FC204 Fraction Collectors
Brand:  Gilson™ 130412
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16626115
  • £804.54 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • HPLC Purification Technique
  • Maximum Vessel Quantity is 80
  • Tube Vessel Type
Specifications
Specifications
Code 4
5 mL Vial (12 x 75 mm)
80