Learn More
Gilson™ Position Rack Tube Vessel Type, Fraction Collector
These racks are for use with the FC203B and FC204 Fraction Collectors
Brand:  Gilson™ 12040101
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16676105
  • £31.43 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • HPLC Purification Technique
  • Maximum Vessel Quantity is 80
  • Tube Vessel Type
Specifications
Specifications
Fraction Collector Racks
Code 1
9 mL Vial (13 x 100 mm)
80