Learn More
Ohaus™ Post Extension Kit
OHAUS™ Post Extension Kit suitable for use with Multi-Tube Vortex Mixers.
Brand: Ohaus™ 30400244
161.50 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.90000kg
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Post Extension Kit | |
Post Extension Kit | |
Multi-Tube Vortex Mixer |