missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Posts and Rubber Bases

Posts and Rubber Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.

Brand:  Brady™ 256723

96.90 GBP valid until 2024-12-31
Use promo code "21877" to get your promotional price.


 View more versions of this product

Product Code. 11721306

  • £103.00 / 1 pack
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Post and Fillable Base.
Specifications

Specifications

POST & RUBBER BASE RED/WHT PK 4
35.43 in.
4 Pk.
Post with Rubber Base
40 mm
900 mm
Red on White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.