missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Post Top
This Post Top is ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
£6.80
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
11786675
|
Brady™
292247 |
Black |
£6.80
Each |
In Stock
Log in to see stock. |
||||||
11796675
|
Brady™
292248 |
Red |
£6.80
Each |
In Stock
Log in to see stock. |
||||||
11706685
|
Brady™
292249 |
White |
£6.80
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||||
Description
- Ideal for controlling and directing foot traffic.
- Suitable for warehouses, factories, offices etc.
- Chains and ribbons can be attached to the sturdy 900 mm posts.
- Top only.
Specifications
60 mm | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Post Top