missing translation for 'onlineSavingsMsg'
Learn More

Notrax Posture Mat™ Anti-Fatigue Mat for Sit-Stand Desks

The Posture Mat™ Anti-Fatigue Mat for Sit-Stand Desks is an exceptionally soft and comfortable anti-fatigue mat for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations.

Brand:  Notrax 426S7052GY

 View more versions of this product

Product Code. 17702498

  • £58.60 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations
  • The rebound of the foam results in an energizing bounce effect that stimulates automatic balance checks to improve posture and constant micro movements to maintain blood flow
  • The ideal anti-fatigue mat for adjustable desks that combines style and comfort primarily for stationary positions making it ideal for sit-stand desks and standing workstations
  • A built-in handle makes it easy to carry or hang up when not in use
Specifications

Specifications

Gray
70 cm
Industrial and Commercial Matting
2.3 ft.
PVC
52 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.