missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Posture Mat Classic™ Anti-fatigue Mats for Standing Workers
Notrax Posture Mat - Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations.
£53.40 - £237.00
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17732128
|
Notrax
425S2024BL |
60 cm |
£53.40
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
17752128
|
Notrax
425S3640BL |
102 cm |
£163.00
Each |
In Stock
Log in to see stock. |
|||||
17762128
|
Notrax
425S3660BL |
152 cm |
£237.00
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
17742128
|
Notrax
425S2472BL |
177 cm |
£198.00
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
- Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations
- The rebound of the foam results in an energizing bounce effect that stimulates automatic balance checks to improve posture and constant micro movements to maintain blood flow
- Made from a durable closed cell foam that is reputed for its ultimate comfort and longevity properties, the naturally resilient material moulds to the foot to spread the weight and reduce pressure points from the hard floor underneath
- All four sides have bevelled edges to minimize tripping hazards
- Loose lay with no installation required
Specifications
Black | |
Rubber | |
Industrial and Commercial Matting |