missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-(azidomethyl)phenyltrifluoroborate, 95%, Thermo Scientific™

Catalog Number p-7039339
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
898544-48-4
C7H6BF3KN3
239.05
MFCD11052751
AKJNDNIFSIXTBR-UHFFFAOYSA-N
potassium 2-azidomethyl phenyltrifluoroborate, potassium 2-azidomethyl phenyl trifluoroborate, potassium 2-azidomethyl phenyl trifluoroboranuide, trifluoro 2-azidomethyl phenyl potassioboron v, potassium 2-azidomethyl phenyl trifluoro borate 1-
57416898
potassium;[2-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=CC=C1CN=[N+]=[N-])(F)(F)F.[K+]
This item is not returnable. View return policy
898544-48-4
C7H6BF3KN3
239.05
MFCD11052751
AKJNDNIFSIXTBR-UHFFFAOYSA-N
potassium 2-azidomethyl phenyltrifluoroborate, potassium 2-azidomethyl phenyl trifluoroborate, potassium 2-azidomethyl phenyl trifluoroboranuide, trifluoro 2-azidomethyl phenyl potassioboron v, potassium 2-azidomethyl phenyl trifluoro borate 1-
57416898
potassium;[2-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=CC=C1CN=[N+]=[N-])(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 241.06
Percent Purity 95%
Chemical Name or Material Potassium 2-(azidomethyl)phenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'