missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-methoxypyridine-5-trifluoroborate, 97%, Thermo Scientific™

Product Code. 15438208
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1144573-61-4
Molecular Formula C6H6BF3KNO
Molecular Weight (g/mol) 215.024
MDL Number MFCD09993111
InChI Key ASWXYOAUUZSFCK-UHFFFAOYSA-N
Synonym potassium 2-methoxypyridine-5-trifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl borate, amtb168, potassium 6-methoxy-3-pyridyltrifluoroborate, potassium trifluoro 6-methoxypyridin-3-yl boranuide
PubChem CID 72698470
IUPAC Name potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide
SMILES [B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]

Specifications

Quantity 1 g
Formula Weight 215.02
Percent Purity 97%
Chemical Name or Material Potassium 2-methoxypyridine-5-trifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'