missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-fluorophenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15409197
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 267006-24-6
Molecular Formula C6H4BF4K
Molecular Weight (g/mol) 202
MDL Number MFCD06659938
InChI Key LOPFKHUYBDOHRD-UHFFFAOYSA-N
Synonym potassium 3-fluorophenyltrifluoroborate, potassium trifluoro 3-fluorophenyl borate, potassium trifluoro 3-fluorophenyl boranuide, potassium 3-fluorophenyl trifluoroborate, potassium trifluoro 3-fluorophenyl borate 1-, pubchem11392, c6h4bf4.k, potassium trifluoro-3-fluorophenyl boranuide, potassium ion trifluoro 3-fluorophenyl boranuide
PubChem CID 23697350
IUPAC Name potassium;trifluoro-(3-fluorophenyl)boranuide
SMILES [B-](C1=CC(=CC=C1)F)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 202
Percent Purity 96%
Chemical Name or Material Potassium 3-fluorophenyltrifluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.