missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15469955
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850623-45-9
Molecular Formula C14H12BF3KNO2
Molecular Weight (g/mol) 333.159
MDL Number MFCD04115743
InChI Key ADBHDDIXRRMUNK-UHFFFAOYSA-N
Synonym potassium 4-cbz-aminophenyl trifluoroborate, potassium 4-benzyloxycarbonylamino phenyltrifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoroboranuide, potassium 4-carboxybenzoylaminophenyl trifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoro borate 1-
PubChem CID 24851538
IUPAC Name potassium;trifluoro-[4-(phenylmethoxycarbonylamino)phenyl]boranuide
SMILES [B-](C1=CC=C(C=C1)NC(=O)OCC2=CC=CC=C2)(F)(F)F.[K+]

Specifications

Melting Point 250°C to 266°C
Quantity 5 g
Formula Weight 333.16
Percent Purity 96%
Chemical Name or Material Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'