missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Plastic Bottle in Tin Can
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Assay Percent Range | 98% |
Linear Formula | K |
Quantity | 500 g |
Fieser | 01,905; 06,274; 12,400; 13,170; 14,308; 15,269 |
Merck Index | 15, 7725 |
Formula Weight | 39.1 |
Packaging | Plastic Drum |
Form | Chunk |
Chemical Name or Material | Potassium |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467815 | Certificate of Analysis | 31/01/2025 | 223860500,223861000,223860100,223865000 |
A0463550 | Certificate of Analysis | 21/06/2024 | 223860500,223861000,223860100,223865000 |
A0458916 | Certificate of Analysis | 21/12/2023 | 223860500,223861000,223860100,223865000 |
A0453046 | Certificate of Analysis | 16/05/2023 | 223860500,223861000,223860100,223865000 |
A0449214 | Certificate of Analysis | 09/01/2023 | 223860500,223861000,223860100,223865000 |
Safety and Handling


Product Identifier
- Potassium, cubes, stored in mineral oil
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- MIXTURE LIST-Contains : potassium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction