missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™

Product Code. 15475119
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Color Colorless
Linear Formula CH3CO2K
Concentration 1M aq. soln.
Formula Weight 98.15
pH 7.5
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Potassium acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'