missing translation for 'onlineSavingsMsg'
Learn More
Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals Product Code.: 11356798

Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals

Product Code. 11356798
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
5 x 2 kg
This item is not returnable. View return policy

Product Code. 11356798

Brand: Thermo Scientific Alfa Aesar 013449.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920Show MoreShow Less
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% min
Beilstein 3595449
Solubility Information Soluble in water,alcohol and ammonia. Insoluble in acetone and ether.
Formula Weight 98.15
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.57 g/mL
Chemical Name or Material Potassium acetate, For ACS analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP19L040 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number013449.36,013449.A3,013449.F7,013449.22
Lot NumberP18L046 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number013449.36,013449.A3,013449.F7,013449.22
Lot NumberP10L016 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog Number013449.36,013449.A3,013449.F7,013449.22
Lot NumberZ19K027 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number013449.36,013449.A3,013449.F7,013449.22
Lot NumberM09L033 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number013449.36,013449.A3,013449.F7,013449.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.