missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium acetate, ACS Reagent, ≥99.0%, Honeywell Fluka™
Click to view available options
25 kg
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
Specifications
Specifications
CAS | 127-08-2 |
Molecular Formula | C2H3KO2 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
SMILES | [K+].CC([O-])=O |
IUPAC Name | potassium acetate |
Molecular Weight (g/mol) | 98.14 |
Quantity | 25 kg |
Chemical Name or Material | Potassium acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'