missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Granular, U.S.P., J.T. Baker™
£189.00 - £761.00
Chemical Identifiers
CAS | 127-08-2 |
---|---|
Molecular Formula | C2H3KO2 |
Molecular Weight (g/mol) | 98.142 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
IUPAC Name | potassium;acetate |
SMILES | CC(=O)[O-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13519040
|
Avantor J.T.Baker
2914-01 |
500 g | Poly Bottle |
£189.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578064
|
Avantor J.T.Baker
2914-05 |
2.5 kg | Poly Bottle | N/A | |||||
Chemical Identifiers
127-08-2 | |
98.142 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
C2H3KO2 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
Specifications
127-08-2 | |
C2H3KO2 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
1L = 1.57kg |
Granules | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
USP | |
Potassium Acetate |