missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate (Molecular Biology), MP Biomedicals
Click to view available options
Quantity:
500 g
Specifications
CAS | 127-08-2 |
Quantity | 500 g |
Molecular Formula | C2H3KO2 |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
SMILES | CC(=O)[O-].[K+] |
IUPAC Name | potassium;acetate |
Molecular Weight (g/mol) | 98.142 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'