missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Specifications
Quantity | 250 g |
Linear Formula | CH3COOK |
UN Number | NONH for all modes of transport |
Beilstein | 3595449 |
Formula Weight | 98.14g/mol |
Percent Purity | ≥99% (perchloric acid titration) |
Grade | puriss. p.a. |
Density | 1.57g/cm3at 25°C |
Chemical Name or Material | Potassium Acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'