missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonyl tartrate sesquihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10772841
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 333.93
MDL Number MFCD00167056
Synonym Antimonyl potassium tartrate sesquihydrate

Specifications

Melting Point 100.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99% min. (Iodometry)
Merck Index 15, 691
Solubility Information Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol
Formula Weight 333.93
Percent Purity ≥99%
Infrared Spectrum Authentic
Loss on Drying 2.7% max. (105°C)
Packaging Glass bottle
Chemical Name or Material Potassium antimonyl tartrate sesquihydrate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'