missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, ≥99.95% Trace Metals Basis, Honeywell™

Product Code. 15665510
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle

≥99.95% trace metals basis, 99.7-100.5% dry basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 100°C to 120°C
Color White
pH 8 to 8.6 (at 20°C)
Quantity 250 g
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Solubility Information In water 333g/L at 20°C
Formula Weight 100.12g/mol
Percent Purity ≥99.95% (trace metals basis)
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 2.17g/cm3 at 20°C
Chemical Name or Material Potassium bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.