missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(oxalato)oxotitanate(IV) dihydrate, Thermo Scientific Chemicals

Product Code. 11433800
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Mordant in dyeing, reagent

Mordant in dyeing, reagent Potassium bis(oxalato)oxotitanate(IV) dihydrate is used as a mordant in dyeing and acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14402-67-6
Molecular Formula C4K2O9Ti
Molecular Weight (g/mol) 318.10
MDL Number MFCD00150592
InChI Key UHWHMHPXHWHWPX-UHFFFAOYSA-J
Synonym Titanium potassium oxalate
IUPAC Name dipotassium oxotitaniumbis(ylium) dioxalate
SMILES [K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Color Colorless
Odor Odorless
Quantity 2 kg
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 354.10 (318.10 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.