missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bisulfate, 99%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 214.0°C |
Quantity | 10 kg |
Assay Percent Range | 99% |
Linear Formula | KHSO4 |
Merck Index | 15,7734 |
Solubility Information | (10% in water) Clear colorless |
IUPAC Name | potassium hydrogen sulfate |
Formula Weight | 136.16 |
Percent Purity | 99% |
Grade | Extra Pure |
Show More |
Safety and Handling
Product Identifier
- Potassium bisulfate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'