missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, 0.033M, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11454799
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.2.2) and prepared in an ISO 9001 facility. Potassium Bromate, 0.033M, EP Grade, Reagecon™ is a ready-to-use volumetric solution with 5.511g/L KBrO3 in water. It is an oxidizing agent used as a laboratory reagent.

Potassium bromate is produced when bromine is passed through a hot solution of potassium hydroxide. This solution is prepared by mixing 5.5670g of potassium bromate primary standard with water and diluting it to 1L. This yields a 0.1M solution with about 5.511g/L KBrO3.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.2.2) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise volumetric solutions

Specifications

CAS 7758-01-2, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula BrKO3
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
Solubility Information Soluble in water
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
ChEBI CHEBI:38211
Formula Weight 167.00g/mol
Grade Eu. Ph. Chapter 4(4.2.2)
Concentration or Composition (by Analyte or Components) 5.5670g of potassium bromate primary standard solution, diluted to 1L with water
Odor Odorless
Packaging Tamper evident
Chemical Name or Material Potassium bromate
Show More Show Less
compliance-icons
Product Identifier
  • 0.033M POTASSIUM BROMATE
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'