missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromate, Certified AR for Analysis, Fisher Chemical™
£161.00 - £420.00
Chemical Identifiers
CAS | 7758-01-2 |
---|---|
Molecular Formula | BrKO3 |
Molecular Weight (g/mol) | 167.00 |
MDL Number | 11359 |
InChI Key | OCATYIAKPYKMPG-UHFFFAOYSA-M |
Synonym | potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromatShow More |
PubChem CID | 23673461 |
ChEBI | CHEBI:38211 |
IUPAC Name | potassium;bromate |
SMILES | [O-]Br(=O)=O.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10704961
|
Fisher Chemical
P/3920/50 |
250 g | N/A | ||||||
10675882
|
Fisher Chemical
P/3920/53 |
500 g | N/A | ||||||
10224300
|
Fisher Chemical
P/3920/60 |
1 kg |
£420.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7758-01-2 | |
167.00 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
BrKO3 | |
11359 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromatShow More | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
Specifications
350°C | |
White | |
Powder/Solid | |
11359 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01g/mol |
7758-01-2 | |
5 to 9 | |
BrKO3 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
[O-]Br(=O)=O.[K+] | |
167.00 | |
CHEBI:38211 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium bromate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Acute toxicity Category 3
- Carcinogenicity Category 1B
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Bromate, Certified AR for Analysis, Fisher Chemical™