missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Potassium bromide is used in a variety of applications such as spectroscopy, infrared transmission, as a laboratory reagent, and for the removal of peripheral membrane proteins. It is also used in photosensitive chemicals, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1435°C |
Melting Point | 730°C |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Linear Formula | KBr |
Merck Index | 14,7618 |
Solubility Information | Soluble in water. Slightly soluble in alcohol,glycerol and ether. |
IUPAC Name | potassium;bromide |
Formula Weight | 119.01 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'