missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, 98%, extra pure, anhydrous, Thermo Scientific Chemicals

£30.35 - £455.27

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15530554
View Documents Promotion Details
Thermo Scientific Acros
447620010
1 kg
£30.35
1kg
Please to purchase this item. Need a web account? Register with us today!
15540554
View Documents
Thermo Scientific Acros
447620050
5 kg
£99.90
5kg
Please to purchase this item. Need a web account? Register with us today!
15550554
View Documents
Thermo Scientific Acros
447620250
25 kg
£455.27
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
[K+].[K+].[O-]C([O-])=O
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
dipotassium;carbonate

Specifications

891.0°C
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
98%
2.0% max.
584-08-7
K2CO3
15,774
Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
[K+].[K+].[O-]C([O-])=O
138.21
138.21
Extra Pure
Potassium carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-529-3

Product Identifier
  • Potassium carbonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Carbonic acid, dipotassium salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only